Learn More
Chemglass Life Sciences 0.2mL PCR 8-Strip Tubes, Clear, 125/Pk
SDP
Catalog No. 501897738
Encompass_Preferred
USP grade VI PolypropyleneLow profile tubes are designed to minimize the condensate contamination, particularly for overnight experiments, and also reduce the evaporation of the reaction solution.Free of Dnase and Rnase, human DNA and PCR inhibitorAutoclavable
Catalog No. 50-189-7738 Supplier Chemglass Life Sciences Supplier No. CGN3610C01
USP grade VI PolypropyleneLow profile tubes are designed to minimize the condensate contamination, particularly for overnight experiments, and also reduce the evaporation of the reaction solution.Free of Dnase and Rnase, human DNA and PCR inhibitorAutoclavable