Learn More
Chemglass Life Sciences 0.2mL PCR 8-Strip Tubes, White, 125/Pk
SDP
Catalog No. 501897739
Encompass_Preferred
USP grade VI PolypropyleneLow profile tubes are designed to minimize the condensate contamination, particularly for overnight experiments, and also reduce the evaporation of the reaction solution.Free of Dnase and Rnase, human DNA and PCR inhibitorAutoclavableWhite strip tubes increase the fluorescent signal strength of real-time PCR
Catalog No. 50-189-7739 Supplier Chemglass Life Sciences Supplier No. CGN3610W01
USP grade VI PolypropyleneLow profile tubes are designed to minimize the condensate contamination, particularly for overnight experiments, and also reduce the evaporation of the reaction solution.Free of Dnase and Rnase, human DNA and PCR inhibitorAutoclavableWhite strip tubes increase the fluorescent signal strength of real-time PCR