Learn More
MTC Bio PCR Plates, Standard 96 well x 0.2ml, non-skirted,WHITE, 50/pk
Catalog No. NC2017824
Encompass_Preferred
Pure•Amp™ PCR Plates are designed to fit all popular thermal cyclers and qPCR instruments. Six configurations (0.1mL, 0.2mL skirted, semi-skirted and without skirt) are available to accommodate equipment from all major manufacturers. In qPCR applications, use white plates to enhance signal strength. All Pure•Amp™ tubes, strips and plates are made of medical grade heat-resistant polypropylene. All Pure•Amp™ products are PCR certified and are guaranteed to be free of RNase, DNase, DNA, pyrogens, endotoxins and PCR inhibitors.
Catalog No. NC2017824 Supplier MTC Bio Supplier No. P9602NW
Pure•Amp™ PCR Plates are designed to fit all popular thermal cyclers and qPCR instruments. Six configurations (0.1mL, 0.2mL skirted, semi-skirted and without skirt) are available to accommodate equipment from all major manufacturers. In qPCR applications, use white plates to enhance signal strength. All Pure•Amp™ tubes, strips and plates are made of medical grade heat-resistant polypropylene. All Pure•Amp™ products are PCR certified and are guaranteed to be free of RNase, DNase, DNA, pyrogens, endotoxins and PCR inhibitors.