Learn More
Bioland Scientific LLC 0.2ml Standard Profile 96-well PCR Plate (non-skirt), 50/cs
SDP
Catalog No. NC1540131
Encompass_Preferred
Non-Skirted. Standard Profile : Tube height 21 mm. Clear Tubes. Thin Walled tube for best heat transfer. Made of USP VI Polypropylene. Free of DNase, RNase, DNA, PCR Inhibitor, ATP, Endotoxin. All products are ready to use in qPCR. Evaporation rate below 5%, saving cost on reagents. Low and standard profile with 3 skirted format for almost all qPCR and sequencer machine. Small package can avoid contamination by different researcher.
Catalog No. NC1540131 Supplier Bioland Scientific LLC Supplier No. PPP9602
Non-Skirted. Standard Profile : Tube height 21 mm. Clear Tubes. Thin Walled tube for best heat transfer. Made of USP VI Polypropylene. Free of DNase, RNase, DNA, PCR Inhibitor, ATP, Endotoxin. All products are ready to use in qPCR. Evaporation rate below 5%, saving cost on reagents. Low and standard profile with 3 skirted format for almost all qPCR and sequencer machine. Small package can avoid contamination by different researcher.