Learn More
USA Scientific Inc 0.2 mL TempAssure PCR tube without cap, natural
Catalog No. NC9400410
Encompass
0.2 mL TempAssure PCR tube without cap, natural. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.
Catalog No. NC9400410 Supplier USA Scientific Inc Supplier No. 14020200
0.2 mL TempAssure PCR tube without cap, natural. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.