Learn More
USA Scientific Inc 0.2 mL TempAssure PCR tube, attached optical cap, natural
Catalog No. NC9066601
Encompass
0.2 mL TempAssure PCR tube, attached optically clear flat cap, natural. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.
Catalog No. NC9066601 Supplier USA Scientific Inc Supplier No. 14028120
0.2 mL TempAssure PCR tube, attached optically clear flat cap, natural. DNase, RNase, DNA, PCR inhibitor, and pyrogen free. 1000/bag.