Learn More
Ace Glass, Inc. 0-3 4 TEF-CAP PLUG (GB)
SDP
Catalog No. 50979703
Encompass_Preferred
Replacement Tef-Cap plug for 0-3mm and 0-4mm High Vacuum Easy Action Stopcocks.
Catalog No. 50-979-703 Supplier Ace Glass, Inc. Supplier No. 818943
Replacement Tef-Cap plug for 0-3mm and 0-4mm High Vacuum Easy Action Stopcocks.