Learn More
ACCU-SCOPE™ 0.35x C-Mount Adapter
GREENER_CHOICESDP
Catalog No. 01189301 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Type:
Camera Adapter
Catalog No. Type
01-189-301 Camera Adapter
1 options
Catalog No. 01-189-301 Supplier ACCU-SCOPE™ Supplier No. 00201035
Add to Cart
Add to Cart
Microscope camera 0.35x c-mount adapter for 1/3 in. chip sensors, with adjustable focus. Fits ACCU-SCOPE Models EXI-310, EXC-350, EXC-100. Adapter can be focused to match the focus of the digital image to the image seen through the eyepieces. Ideal for use with 0.35 in. diagonal camera sensors or smaller.
  • 0.35x c-mount adapter to attach camera to EXC-400, EXC-350, 3001-LED or 3001 microscopes.
  • Adapter can be focused to match the focus of the digital image to the image seen through the eyepieces.
  • Ideal for use with 0.35 in. diagonal camera sensors or smaller.
Specifications
Type Camera Adapter
Autoclavable Not Autoclavable
Sterility Non-sterile