Learn More
Motic™ 0.35X C-Mount Camera Adapter
Catalog No. 01258051 Shop All Motic Group Products
Change view
Click to view available options
Type:
Camera Adapter
Catalog No. Type
01-258-051 Camera Adapter
1 options
This item is not returnable. View return policy
Catalog No. 01-258-051 Supplier Motic™ Supplier No. 1101001904111
Add to Cart
Add to Cart
This item is not returnable. View return policy
This C-mount camera adapter allows Moticams to connect with Motic™ trinocular microscopes.
Compatible with the following cameras:
  • Moticam 2
  • Moticam X
  • Moticam X3
  • Moticam 1080
  • Moticam BTU 10
  • Moticam BTX 8
Specifications
Type Camera Adapter
For Use With (Equipment) Motic BA, AE2000/AE31E, SMZ-161/171 Trinocular Microscopes
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov