Learn More
Alkali Scientific 0.3mL 11mm PP Snap Top Micro Vial, 100/Pk, Amber
SDP
Supplier:  Alkali Scientific MVP03AMB
Encompass_Preferred
The Alkali Scientific™ 0.3 mL PP Snap Top Micro Vial is designed for secure storage and precise handling of very small sample volumes in laboratory and analytical applications. Made from high quality polypropylene (PP), this micro vial provides excellent chemical resistance, strength, and durability for routine laboratory workflows. The integrated snap top closure forms a reliable seal without requiring additional caps, helping maintain sample integrity during storage, handling, and transport. Measuring approximately 11 mm in diameter, the compact design supports efficient small volume handling and high throughput laboratory processes. Supplied in convenient packs of 100, these micro vials are suitable for chromatography, analytical testing, and general laboratory procedures. The clear polypropylene construction allows easy visibility of sample contents, making them ideal for chromatography preparation, sample storage, and routine analytical laboratory work.
Catalog No. 50-006-62157