Learn More
Alkali Scientific 0.3mL 9 425 PP Screw Thread Micro Vial Transparent. 12x32mm, 100/Pk, Transparent
Supplier: Alkali Scientific MVP03ST
The Alkali Scientific™ 0.3 mL Micro Vial is designed for precise handling of very small sample volumes in laboratory and analytical applications. Made from high quality transparent polypropylene (PP), this vial offers excellent chemical resistance and clear visibility of contents. The 9 425 screw thread ensures compatibility with standard caps and closures, while the 12 × 32 mm dimensions make it ideal for autosampler and high throughput workflows. Supplied in packs of 100, these micro vials are perfect for chromatography, small volume sample storage, and routine laboratory analysis. Key Features 0.3 mL capacity for precise handling of small volume samples Made from transparent polypropylene (PP) for excellent chemical resistance Clear material allows full visibility of sample contents 9 425 screw thread compatible with standard caps and closures 12 × 32 mm dimensions suitable for autosamplers and high throughput workflows Light and durable for routine laboratory use Ideal for c