Learn More
WELCH MATERIALS INC DoprahTM 0.3mL 9-425 PP Vial: 0.3mL 9-425 PP Screw Thread Micro-Vial Transparent. 12x32mm. 100pcs/pk, 10pk/cs.
Catalog No. NC2053886
Encompass_Preferred
0.3mL 9-425 PP Vial: 0.3mL 9-425 PP Screw Thread Micro-Vial Transparent. 12x32mm. 100pcs/pk, 10pk/cs.
Catalog No. NC2053886 Supplier WELCH MATERIALS INC Supplier No. DQ118410
0.3mL 9-425 PP Vial: 0.3mL 9-425 PP Screw Thread Micro-Vial Transparent. 12x32mm. 100pcs/pk, 10pk/cs.