Learn More
Glass Expansion 0.3mm ID PTFE Sheathed Carbon Fiber Probe with 686mm long capillary and UniFit Connector (for Agilent I-AS Autosamplers)
SDP
Catalog No. 502703094
Encompass_Preferred
PTFE Sheathed Carbon Fiber Probe 0.3mm ID with 686mm long capillary and UniFit Connector. Suitable for Agilent I-AS Autosamplers. UniFit Connector attaches directly to Glass Expansion U-Series and DC Nebulizers.Replacement of autosampler probe is recommended about every 6 months to a year. This will ensure you have unrestricted sample flow. If clogs within probes or nebulizers occur, consider using the Guardian In-Line Sample Filter (P/N NC0897716, 70-803-1108). The Guardian In-Line Sample Filter is spliced into the autosampler probe capillary & can be backflushed using the Eluo Nebulizer Cleaner for Glass Concentric Nebulizers (NC9462559, 70-ELUO), or the Eluo HF Nebulizer Cleaner for OpalMist or DuraMist (501907353, 70-ELUO-OPD), & Eluo Adaptor for In-Line Filter (NC0801128, 70-803-1160). The Guardian In-Line Sample Filter is reuseable & can last years.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3094 Supplier Glass Expansion Supplier No. 607031009
PTFE Sheathed Carbon Fiber Probe 0.3mm ID with 686mm long capillary and UniFit Connector. Suitable for Agilent I-AS Autosamplers. UniFit Connector attaches directly to Glass Expansion U-Series and DC Nebulizers.Replacement of autosampler probe is recommended about every 6 months to a year. This will ensure you have unrestricted sample flow. If clogs within probes or nebulizers occur, consider using the Guardian In-Line Sample Filter (P/N NC0897716, 70-803-1108). The Guardian In-Line Sample Filter is spliced into the autosampler probe capillary & can be backflushed using the Eluo Nebulizer Cleaner for Glass Concentric Nebulizers (NC9462559, 70-ELUO), or the Eluo HF Nebulizer Cleaner for OpalMist or DuraMist (501907353, 70-ELUO-OPD), & Eluo Adaptor for In-Line Filter (NC0801128, 70-803-1160). The Guardian In-Line Sample Filter is reuseable & can last years.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001