Learn More
Chemglass Life Sciences Adapter, Connecting, Airfree, Schlenk, 0-4mm Valve
SDP
Catalog No. AF007601
Encompass_Preferred
Adapter, Connecting, Airfree, Schlenk, 0-4mm Valve. Adapter has two #15 o-ring joints connected in-line via a Chem-Cap valve. Available with either 0-4mm or 0-8mm valve sizes. Each adapter is supplied complete with two #116 Viton o-rings.
Catalog No. AF007601 Supplier Chemglass Life Sciences Supplier No. AF007601
Adapter, Connecting, Airfree, Schlenk, 0-4mm Valve. Adapter has two #15 o-ring joints connected in-line via a Chem-Cap valve. Available with either 0-4mm or 0-8mm valve sizes. Each adapter is supplied complete with two #116 Viton o-rings.