Learn More
Ace Glass, Inc. Gauge, only, pressure, 0-400psi, brass internal materials, 1/4inmale NPT bottom fitting, 2.5in diameter
SDP
Catalog No. 50977963
Encompass
0-400psi pressure gauge with brass internal materials. Features male NPT bottom fitting. Gauges diameter is 2.5 inches.
Catalog No. 50-977-963 Supplier Ace Glass, Inc. Supplier No. 1338512
0-400psi pressure gauge with brass internal materials. Features male NPT bottom fitting. Gauges diameter is 2.5 inches.