Learn More
Chemglass Life Sciences 0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum
SDP
Catalog No. CG96101
Encompass_Preferred
0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum. Replacement CHEM-CAP plug and control knob for CHEM-VAC high vacuum valves CG-951 through CG-960. Plug is sold complete with control knob and CHEM-CAP stem.
Catalog No. CG96101 Supplier Chemglass Life Sciences Supplier No. CG96101
0-4mm Chem-Vac, Chem-Cap Valve Replacement Plug and Control Knob, High Vacuum. Replacement CHEM-CAP plug and control knob for CHEM-VAC high vacuum valves CG-951 through CG-960. Plug is sold complete with control knob and CHEM-CAP stem.