Learn More
Andwin Scientific 0.45UM GHP ACRODISC LC25 50/PK
SDP
Supplier:  Andwin Scientific 4560PK
Encompass
The 0.45 �m Acrodisc� syringe filter with GHP (hydrophilic polypropylene) membrane feature low protein binding. The GHP membrane is also HPLC certified for low levels of UV-absorbing extractables. This product is sold in a pack of 50 filters.
Catalog No. NC0244104
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.