Learn More
Chemglass Life Sciences 0-4mm Glass Plug with Control Knob, Viton O-Rings
SDP
Catalog No. 501531879
Encompass_Preferred
0-4mm Glass Plug with Control Knob, Viton O-Rings. Replacement glass plug and control knob for CG-973 through CG-982 high vacuum valves. Plug is sold complete with control knob and glass stem having three o-rings. Available with a choice of Viton or Kalrez o-rings.
Catalog No. 50-153-1879 Supplier Chemglass Life Sciences Supplier No. CG98301
0-4mm Glass Plug with Control Knob, Viton O-Rings. Replacement glass plug and control knob for CG-973 through CG-982 high vacuum valves. Plug is sold complete with control knob and glass stem having three o-rings. Available with a choice of Viton or Kalrez o-rings.