Learn More
Chemglass Life Sciences Valve, 0-4mm, In-Line, Glass Only
SDP
Catalog No. 501532018
Encompass_Preferred
Valve, 0-4mm, In-Line, Glass Only. Hi-vac valve with bent arms to minimize hold up of fluids that would occur in conventional valves. Designed with a precision external thread for fine control.
Catalog No. 50-153-2018 Supplier Chemglass Life Sciences Supplier No. CGK50501
Valve, 0-4mm, In-Line, Glass Only. Hi-vac valve with bent arms to minimize hold up of fluids that would occur in conventional valves. Designed with a precision external thread for fine control.