Learn More
Waters Corp Microtube Domino, 0.5-0.6 mL, 24 Microtubes, Machine-Readable
Catalog No. NC3717741 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 0.5-0.6 mL Microtube Domino is designed to hold small-volume microtubes. Its design maintains attached caps of certain tubes in a vertical position to avoid automation collisions. It can hold up to 24 microtubes. Dominos are machine-readable consumable holders that interface various consumable types with the Andrew+ pipetting robot for seamless automation of laboratory workflows.
  • Designed to hold small-volume microtubes.
  • Maintains attached caps of certain tubes in a vertical position to avoid automation collisions.
  • Can hold up to 24 microtubes.
  • Machine-readable consumable holders create the interface between different consumable types and a pipetting robot.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.
Catalog No. NC3717741 Supplier Waters Corp Supplier No. 186010096
The 0.5-0.6 mL Microtube Domino is designed to hold small-volume microtubes. Its design maintains attached caps of certain tubes in a vertical position to avoid automation collisions. It can hold up to 24 microtubes. Dominos are machine-readable consumable holders that interface various consumable types with the Andrew+ pipetting robot for seamless automation of laboratory workflows.
  • Designed to hold small-volume microtubes.
  • Maintains attached caps of certain tubes in a vertical position to avoid automation collisions.
  • Can hold up to 24 microtubes.
  • Machine-readable consumable holders create the interface between different consumable types and a pipetting robot.
  • Equipped with QR codes for critical labware information.
  • Equipped with ARUCO codes for spatial positioning.