Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab 0.5% Focus Acid Alcohol
SDP

Catalog No. 22000079
Change view
Click to view available options
Quantity:
Gallon
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-000-079 Gallon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-079 Supplier StatLab Supplier No. SL881
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Contains hydrochloric acid in 70% reagent grade alcohol

  • Used to remove excess stain and to produce better defined nuclei
  • For section differentiation as a part of regressive staining methods
  • Acid alcohol is used for monochromatic and polychromatic staining methods

Specifications

Physical Form Solution
CAS 64-17-5, 7732-18-5, 67-63-0, 67-56-1, 7647-01-0
Color Clear Colorless
pH 1.5
Quantity Gallon
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.