Learn More
Rockland Immunochemicals 0.5 M EDTA pH 8.0 - MB-014
SDP
Catalog No. 501058978
Encompass_Preferred
EDTA is widely used for scavenging metal ion in biochemistry and molecular biology. Ion depletion is commonly used to deactivate metal-dependent enzymes, either as an assay for their reactivity or to suppress damage to DNA or proteins. In tissue culture EDTA is used as a chelating agent that binds to calcium and prevents joining of cadherins between cells, preventing clumping of cells grown in liquid suspension, or detaching adherent cells for passaging. EDTA is also known to inhibit a range of metallopeptidases, the method of inhibition occurs via the chelation of the metal ion required for catalytic activity.
Catalog No. 50-105-8978 Supplier Rockland Immunochemicals Supplier No. MB014
EDTA is widely used for scavenging metal ion in biochemistry and molecular biology. Ion depletion is commonly used to deactivate metal-dependent enzymes, either as an assay for their reactivity or to suppress damage to DNA or proteins. In tissue culture EDTA is used as a chelating agent that binds to calcium and prevents joining of cadherins between cells, preventing clumping of cells grown in liquid suspension, or detaching adherent cells for passaging. EDTA is also known to inhibit a range of metallopeptidases, the method of inhibition occurs via the chelation of the metal ion required for catalytic activity.