Learn More
Inorganic Ventures 0.5 M Sodium Carbonate Eluent
SDP
Catalog No. 50723173
Encompass_Preferred
Inorganic Ventures' 0.5 M Sodium Carbonate Eluent is a 100mL reference material set in a water matrix for stability. This eluent is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-173 Supplier Inorganic Ventures Supplier No. CARB100ML
Inorganic Ventures' 0.5 M Sodium Carbonate Eluent is a 100mL reference material set in a water matrix for stability. This eluent is Traceable to NIST and manufactured in accordance with our stringent Quality Assurance guidelines. See Certificate of Analysis for expiration.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search