Learn More
Alkali Scientific 0.5 mL 96-Well Deep Well Plate, V-Bottom, Round Well, Sterile, 10/pk, 50/cs
SDP
Supplier:  Alkali Scientific MPD31S
Encompass_Preferred
Virgin, heat-resistant, medical-grade polypropylene plates. RNase, DNase & pyrogen free. Minimal binding & liquid retention. Black printed alpha-numeric reference grids. V (conical) bottoms. Dimensions: 5" L x 3.35" W x 0.57" Hgt. Autoclavable. Temperature range: -86°C to 82.2°C.
Catalog No. 50-238-8924