Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial 0.5 mL, Self-Standing, External Thread, Sterile (CGN-4762-001)
SDP
Catalog No. 502733394
Encompass_Preferred
Chemglass cryogenic vial with 0.5 mL capacity designed for reliable cryogenic storage and sample handling. The vial features a self-standing round bottom and external thread for secure closure, and is manufactured from USP Class VI polypropylene in a controlled cleanroom environment. It is sterilized by e-beam, non-pyrogenic, and suitable for storage in the gas phase of liquid nitrogen.
  • Self-standing round bottom for stable upright storage.
  • External thread cap design for secure closure.
  • Sterile; sterilized by e-beam for immediate use.
  • USP Class VI polypropylene construction for biocompatibility.
  • Clear graduations and white marking area for sample identification.
  • Non-pyrogenic, DNase/RNase free to protect samples.
  • Packaged in 9 x 9 racks; 16 racks per case (1,296 vials per case).
  • Autoclavable; recommended for storage in the gas phase of nitrogen tanks.
Catalog No. 50-273-3394 Supplier Chemglass Life Sciences Supplier No. CGN4762001
Chemglass cryogenic vial with 0.5 mL capacity designed for reliable cryogenic storage and sample handling. The vial features a self-standing round bottom and external thread for secure closure, and is manufactured from USP Class VI polypropylene in a controlled cleanroom environment. It is sterilized by e-beam, non-pyrogenic, and suitable for storage in the gas phase of liquid nitrogen.
  • Self-standing round bottom for stable upright storage.
  • External thread cap design for secure closure.
  • Sterile; sterilized by e-beam for immediate use.
  • USP Class VI polypropylene construction for biocompatibility.
  • Clear graduations and white marking area for sample identification.
  • Non-pyrogenic, DNase/RNase free to protect samples.
  • Packaged in 9 x 9 racks; 16 racks per case (1,296 vials per case).
  • Autoclavable; recommended for storage in the gas phase of nitrogen tanks.