Learn More
Andwin Scientific 0.5 ML SEAL-RITE MICROCENTRIF
SDP
Catalog No. 50819843
Encompass_Preferred
The 0.5 mL Seal-Rite microcentrifuge tube offers secure sample storage with a reliable and precise sealing system that eliminates the chances of accidental leakages and openings. This product is free of RNase, DNAse, DNA, pyrogen, and processing additives like slip agents, biocides, or antistatic agents. The side of the tube is also frosted, allowing users to write on it and easily identify the sample inside.
Catalog No. 50-819-843 Supplier Andwin Scientific Supplier No. 16050005
The 0.5 mL Seal-Rite microcentrifuge tube offers secure sample storage with a reliable and precise sealing system that eliminates the chances of accidental leakages and openings. This product is free of RNase, DNAse, DNA, pyrogen, and processing additives like slip agents, biocides, or antistatic agents. The side of the tube is also frosted, allowing users to write on it and easily identify the sample inside.