Learn More
BioSpec Products .5 MM Glass Beads, 1LB.
SDP
Catalog No. NC0417355
Encompass_Preferred
Glass has a density of 2.5 g/cc and is the most commonly used bead media for "beadbeating."  Beads are sold in one pound bottles with easy pour spouts.  Listed bead sizes are median (average) values.  Because beads are sorted by sieves, bead size varies between +/-10% of a listed value.
Catalog No. NC0417355 Supplier BioSpec Products Supplier No. 11079105
Glass has a density of 2.5 g/cc and is the most commonly used bead media for "beadbeating."  Beads are sold in one pound bottles with easy pour spouts.  Listed bead sizes are median (average) values.  Because beads are sorted by sieves, bead size varies between +/-10% of a listed value.