Learn More
BioSpec Products 0.5 mm Zirconia/Silica Beads 1 lb bottle
SDP
Catalog No. NC0450473
Encompass_Preferred
Zirconia/Silica has a density of 3.7g/cc (50% more dense than glass - good for spores and most tissues)  Listed bead sizes are median (average) values.  Because beads are sorted by sieves, bead size varies between +/-10% of a listed value 1 lb bottle
Catalog No. NC0450473 Supplier BioSpec Products Supplier No. 11079105Z
Zirconia/Silica has a density of 3.7g/cc (50% more dense than glass - good for spores and most tissues)  Listed bead sizes are median (average) values.  Because beads are sorted by sieves, bead size varies between +/-10% of a listed value 1 lb bottle