Learn More
Teknova 0.5 Sodium Hydroxide, 20L Bag, Sterile. 4 Pack.
SDP
Catalog No. 502745810
Encompass
Sodium hydroxide, also known as lye, is a caustic substance that is commonly used as a base in labs for neutralizing acids or creating an alkaline environment for reactions. It is available in a variety of concentrations based on application needs.
Catalog No. 50-274-5810 Supplier Teknova Supplier No. 213414PK
Sodium hydroxide, also known as lye, is a caustic substance that is commonly used as a base in labs for neutralizing acids or creating an alkaline environment for reactions. It is available in a variety of concentrations based on application needs.