Learn More
Grainger 0.5 W T3-1/4 Miniature Bayonet (BA9s)
Catalog No. 502753064
Encompass_Preferred
The Grainger 14A140 is a 14 gauge, 3 conductor SJOOW portable power cable that is rated for 600 volts. It features an oil resistant thermoplastic elastomer jacket and is designed for use in wet or damp locations. This 15 foot cable has a current carrying capacity of 25 amps and operating temperature range of -31°F to 140°F. It can be used for wiring pumps, lighting, portable power distribution, and other wet location applications requiring a flexible, durable cable. This Made in USA cable meets UL standards 62 and 1072.
Catalog No. 50-275-3064 Supplier Grainger Supplier No. 14A140
The Grainger 14A140 is a 14 gauge, 3 conductor SJOOW portable power cable that is rated for 600 volts. It features an oil resistant thermoplastic elastomer jacket and is designed for use in wet or damp locations. This 15 foot cable has a current carrying capacity of 25 amps and operating temperature range of -31°F to 140°F. It can be used for wiring pumps, lighting, portable power distribution, and other wet location applications requiring a flexible, durable cable. This Made in USA cable meets UL standards 62 and 1072.