Learn More
Perkin Elmer US LLC Spacer for Demountable Cell - PTFE, 0.5 mm, Qty. 3
Catalog No. 509052841
Encompass_Preferred
PerkinElmer supplies round spacers made of either lead or teflon. These spacers come in a variety of thicknesses, allowing you to customize the Universal Demountable Cell's path length to the experiment's requirements.
Catalog No. 50-905-2841 Supplier Perkin Elmer US LLC Supplier No. 51001141
PerkinElmer supplies round spacers made of either lead or teflon. These spacers come in a variety of thicknesses, allowing you to customize the Universal Demountable Cell's path length to the experiment's requirements.