Learn More
Elemental Scientific Inc 0.51 PVC FLARED TUBING, ORANGE
SDP
Catalog No. NC2006977
Encompass_Preferred
Peristaltic Pump Tubing, PVC, 3-stop, 0.51mm ID, flared(orange/yellow/orange), 72mm bridge distance, 12 pc.
Catalog No. NC2006977 Supplier Elemental Scientific Inc Supplier No. ES43973051
Peristaltic Pump Tubing, PVC, 3-stop, 0.51mm ID, flared(orange/yellow/orange), 72mm bridge distance, 12 pc.