Learn More
Trajan Scientific 0.5BR-RTC/RSH-5.7/0.63C 0.5UL
SDP
Catalog No. 502530429
Encompass_Preferred
Trajan Autosampler Syringes With Removable Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.024 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.24 in., 57 mm, Model: 0.5BR-RTC/RSH-5.7/0.63C, Volume: 0.5 uL, Connection Type: Removable
Catalog No. 50-253-0429 Supplier Trajan Scientific Supplier No. 000480
Trajan Autosampler Syringes With Removable Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.024 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.24 in., 57 mm, Model: 0.5BR-RTC/RSH-5.7/0.63C, Volume: 0.5 uL, Connection Type: Removable