Learn More
World Precision Instrument Manipulator Mounting Clamp, 1/2-inch
SDP
Catalog No. 50822971
Encompass
Mount your manipulator
  • Diameter: 1/2-inch clamp
  • Interchangeable clamp allows manipulator to be mounted on a variety of supports
Catalog No. 50-822-971 Supplier World Precision Instrument Supplier No. M6
Mount your manipulator
  • Diameter: 1/2-inch clamp
  • Interchangeable clamp allows manipulator to be mounted on a variety of supports