Learn More
Teknova 0.5M HEPES, pH 7.5. 1000mL, Sterile. 2 pack.
SDP
Catalog No. 502387805
Encompass
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.
Catalog No. 50-238-7805 Supplier Teknova Supplier No. H15752PK
HEPES solution is a Goods buffer that is commonly used in cell culture and biochemical applications. It is well known for its ability to maintain pH level, making it an essential part of any experiment that requires a stable pH. It has a buffering range of 6.8 - 8.2.