Learn More
Teknova 0.5M IPTG. 1mL Tubes, 50 Tubes per Pack, Sterile. 2 pack.
SDP
Catalog No. 502745939
Encompass
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a galactose analog that can be used to induce expression of cloned genes under control of the lac operon. It is often used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies.
Catalog No. 50-274-5939 Supplier Teknova Supplier No. I35012PK
IPTG (isopropyl-beta-D-thiogalactopyranoside) is a galactose analog that can be used to induce expression of cloned genes under control of the lac operon. It is often used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies.