Learn More
TRIBIOSCIENCE INC 0.5M TCEP-HCl Solution pH 8.0, 50ML
SDP
Catalog No. NC2398568
Encompass
Tris(2-carboxyethyl) phosphine hydrochloride (TCEP) Solution is an extraordinarily powerful, odorless, thiol-free reducing agent with broad application to protein and nucleic acid involving the reduction of disulfide bonds. The pH value of the solution is designed at 6.0, 7.0, and 8.0 for multiple choices. 
Catalog No. NC2398568 Supplier TRIBIOSCIENCE INC Supplier No. TBS5082508.0
Tris(2-carboxyethyl) phosphine hydrochloride (TCEP) Solution is an extraordinarily powerful, odorless, thiol-free reducing agent with broad application to protein and nucleic acid involving the reduction of disulfide bonds. The pH value of the solution is designed at 6.0, 7.0, and 8.0 for multiple choices. 