Learn More
Hach Company Hach 0.5 mg/L NH4-N Standard Solution for Amtax sc, 2000 mL
Catalog No. NC3659968 Shop All Hach Company Products
Encompass
Ready-to-use 0.5 mg/L NH4-N calibration standard supplied in a 2,000 mL bottle for calibration and verification of online ammonia analyzers such as the Amtax sc. Formulated to provide a nominal ammonium-nitrogen concentration for routine instrument checks and quality control.
  • Ready-to-use calibration standard.
  • Nominal concentration of 0.5 mg/L NH4-N.
  • Supplied in a 2,000 mL bottle for multiple calibrations.
  • Suitable for routine calibration and accuracy verification of online ammonia analyzers.
  • Store and handle according to the product safety data sheet.
Catalog No. NC3659968 Supplier Hach Company Supplier No. 2514654
Ready-to-use 0.5 mg/L NH4-N calibration standard supplied in a 2,000 mL bottle for calibration and verification of online ammonia analyzers such as the Amtax sc. Formulated to provide a nominal ammonium-nitrogen concentration for routine instrument checks and quality control.
  • Ready-to-use calibration standard.
  • Nominal concentration of 0.5 mg/L NH4-N.
  • Supplied in a 2,000 mL bottle for multiple calibrations.
  • Suitable for routine calibration and accuracy verification of online ammonia analyzers.
  • Store and handle according to the product safety data sheet.