Learn More
ANALYTICAL SCIENTIFIC INSTRUMENTS US INC 0.5MICRON TITANIUM FRIT 5/PK
SDP
Catalog No. NC2311923
Encompass
0.5 micron Titanium Frit UltraShield HPLC/UHPLC Precolumn Filter AssemblyPort- Halo compatible / Male- Halo compatible
Catalog No. NC2311923 Supplier ANALYTICAL SCIENTIFIC INSTRUMENTS US INC Supplier No. 8501010T05
0.5 micron Titanium Frit UltraShield HPLC/UHPLC Precolumn Filter AssemblyPort- Halo compatible / Male- Halo compatible