Learn More
Analytical Sales & Services 0.5mL 96-Well Clear Flexi-Tier™ Block System
Catalog No. 5000807278
Encompass
0.5mL 96-Well Clear Flexi-Tier Block System\n\nIncludes:\n• 0.5mL Flat Bottom Glass Inserts\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH
Catalog No. 50-008-07278 Supplier Analytical Sales & Services Supplier No. 96BB05C
0.5mL 96-Well Clear Flexi-Tier Block System\n\nIncludes:\n• 0.5mL Flat Bottom Glass Inserts\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH