Learn More
Analytical Sales & Services 0.5mL 96-Well Clear Flexi-Tier™ Block System
Supplier: Analytical Sales & Services 96FP05C
0.5mL 96-Well Clear Flexi-Tier Block System\n\nIncludes:\n• 0.5mL Conical Glass Inserts\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: Each