Learn More
USA Scientific Inc 0.5 mL deep 96-well PP plate, bulk
Catalog No. NC9977223
Encompass
0.5 mL deep 96-well PlateOne polypropylene plate, round wells, natural, bulk 32/pack
Catalog No. NC9977223 Supplier USA Scientific Inc Supplier No. 18965000
0.5 mL deep 96-well PlateOne polypropylene plate, round wells, natural, bulk 32/pack