Learn More
Analytical Sales & Services 0.5mL 96-Well Opaque Flexi-Tier™ Block System
Catalog No. 5000806110
Encompass
0.5mL 96-Well Opaque Flexi-Tier Block System\n\nIncludes:\n• 0.5mL Conical Glass Inserts\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH
Catalog No. 50-008-06110 Supplier Analytical Sales & Services Supplier No. 96FP05
Add to Cart
Add to Cart
0.5mL 96-Well Opaque Flexi-Tier Block System\n\nIncludes:\n• 0.5mL Conical Glass Inserts\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH