Learn More
Alkali Scientific 0.5mL 96Well, Deep Well Plate, V Conical Bottom, Square Well, Sterile, 50/cs
SDP
Catalog No. 502388671
Encompass_Preferred
Low binding medical grade polypropylene construction. For use with Viral RNA extraction, DNA extraction, RNA/DNA quantification, and immunopreciptitations.
Catalog No. 50-238-8671 Supplier Alkali Scientific Supplier No. MPD22S
Low binding medical grade polypropylene construction. For use with Viral RNA extraction, DNA extraction, RNA/DNA quantification, and immunopreciptitations.