Learn More
Analytical Sales & Services 0.5mL Aluminum Block System, Solid Base with Conical Inserts
Catalog No. 5000806466
Encompass
0.5mL Aluminum Block System with Solid Base. Includes: Aluminum Base, Cover, Screws, Molded Silicone Liner and 0.5mL Clear Glass Conical Inserts Preassembled\n\nQty/Pkg: EACH
Catalog No. 50-008-06466 Supplier Analytical Sales & Services Supplier No. 96A105
0.5mL Aluminum Block System with Solid Base. Includes: Aluminum Base, Cover, Screws, Molded Silicone Liner and 0.5mL Clear Glass Conical Inserts Preassembled\n\nQty/Pkg: EACH