Learn More
Chemglass Life Sciences Chemglass 0.5 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile, Racked (81 per rack, 1,296 per case)
SDP
Catalog No. 502733344
Encompass_Preferred
This 0.5 mL sterile cryogenic vial is designed for long-term sample storage and handling at cryogenic temperatures. It features a self-standing round bottom and an internal thread closure for secure sealing and reliable handling in racked storage.
  • 0.5 mL nominal volume.
  • Self-standing, round-bottom design.
  • Internal thread closure for secure sealing.
  • Sterilized by E-beam.
  • Manufactured from USP 6 grade raw materials.
  • 9 x 9 per rack (81 vials per rack); 16 racks per case (1,296 vials per case).
  • Clear graduations and white marking area.
  • Autoclavable.
  • Non-pyrogenic; DNase/RNase free.
  • Recommended storage in the gas phase of liquid nitrogen.
  • Manufactured in a 100,000 grade cleaning room; ISO 9001 and ISO 13485 certified.
Catalog No. 50-273-3344 Supplier Chemglass Life Sciences Supplier No. CGN4766009
This 0.5 mL sterile cryogenic vial is designed for long-term sample storage and handling at cryogenic temperatures. It features a self-standing round bottom and an internal thread closure for secure sealing and reliable handling in racked storage.
  • 0.5 mL nominal volume.
  • Self-standing, round-bottom design.
  • Internal thread closure for secure sealing.
  • Sterilized by E-beam.
  • Manufactured from USP 6 grade raw materials.
  • 9 x 9 per rack (81 vials per rack); 16 racks per case (1,296 vials per case).
  • Clear graduations and white marking area.
  • Autoclavable.
  • Non-pyrogenic; DNase/RNase free.
  • Recommended storage in the gas phase of liquid nitrogen.
  • Manufactured in a 100,000 grade cleaning room; ISO 9001 and ISO 13485 certified.