Learn More
JDM LAB SOLUTIONS LLC 0.5ml Internal thread 2D Tube
Supplier:  JDM LAB SOLUTIONS LLC 2010031
Encompass
0 5ml 2D barcoded internal thread storage tube precapped with screw cap sterile
Catalog No. 50-000-14297