Learn More
Sarstedt Inc 0.5ML LSC MTUBE CBS ST/PK1000
Catalog No. NC9974441 Shop All Sarstedt Inc. Products
Encompass_Preferred
0.5mL Loop Screw Cap Micro Tube Conical Bottom Skirted O-Ring Cap Assembled Sterile
Catalog No. NC9974441 Supplier Sarstedt Inc Supplier No. 72.730.105
0.5mL Loop Screw Cap Micro Tube Conical Bottom Skirted O-Ring Cap Assembled Sterile