Learn More
Sarstedt Inc 0.5ML LSC MTUBE CBS WB/PK1000
Supplier:  Sarstedt Inc 72.730.106
Encompass_Preferred
0.5mL Loop Screw Cap Micro Tube Conical Bottom Skirted Writing Block O-Ring Cap Assembled Sterile
Catalog No. NC9704437