Learn More
Sarstedt Inc Screw Cap Micro Tube 0.5ML, Conical base with skirted base, 1000/case
Catalog No. NC9216421 Shop All Sarstedt Inc. Products
Encompass_Preferred
Screw Cap Micro Tube, 0.5 ml, PP, with smear edge, with knurls, with conical bottom skirted writing block O-Ring Cap Assembled Sterile, no print, 100 pcs./bag , 10 bags/case, 1000 total
Catalog No. NC9216421 Supplier Sarstedt Inc Supplier No. 72.730.306
Screw Cap Micro Tube, 0.5 ml, PP, with smear edge, with knurls, with conical bottom skirted writing block O-Ring Cap Assembled Sterile, no print, 100 pcs./bag , 10 bags/case, 1000 total